missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD31/PECAM-1 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
251.00 € - 462.00 €
Specifications
| Antigen | CD31/PECAM-1 |
|---|---|
| Clone | 9G11 |
| Dilution | Flow Cytometry 10 μL/10^6 cells, Knockout Validated |
| Applications | Flow Cytometry, Immunoassay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18728589
![]() |
R&D Systems
FAB3567A-025 |
25 Tests |
251.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18758476
![]() |
R&D Systems
FAB3567A-100 |
100 Tests |
462.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human samples. It is validated for Flow Cytometry, Knockout Validated.Specifications
| CD31/PECAM-1 | |
| Flow Cytometry 10 μL/10^6 cells, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 5175 | |
| Activated HUVEC human umbilical vein endothelial cells | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 9G11 | |
| Flow Cytometry, Immunoassay | |
| APC | |
| Mouse | |
| Human | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| PECAM1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human CD31/PECAM-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) E-Selectin, rhICAM-1, -2, -3, rhVCAM-1, or recombinant mouse VCAM-1 is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title