Learn More
Connexin-43 Mouse, Unlabeled, Clone: 2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610062
Description
Gap junctions are intercellular protein pores. They enable cell-to-cell communication by allowing passage of ions and other small molecules. The subunits of gap junction channels are assembled from a family of proteins called connexins. Individual connexin molecules join to make hexameric hemichannels termed connexons; these structures dock to connexons on neighboring cells, forming gap junction pores. Connexin-43 is a member of the connexin family possessing four transmembrane regions, with cytoplasmic amino and carboxyl terminals. It undergoes rapid turnover in the cell and its monomers may reside in the ER/Golgi network, forming a reservoir available for assembly upon
degradation of existing connexin-43 channels. In addition, it is believed that phosphorylation of connexin-43 plays a regulatory role both in the assembly of connexons and in gating activity at the gap junction.
Host Species: Mouse
Clone: 2
Isotype: IgG2
Species Reactivity: Rat
Immunogen: Rat Connexin-43 aa. 252-270
Formula Weight [Chemical]: 43kDa
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| Connexin-43 | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Rat Connexin-43 aa. 252-270 | |
| 150 μg | |
| Cell Biology | |
| Canine, Chicken, Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| 2 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.