missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CXCR7/RDC-1 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
251.00 € - 603.00 €
Specifications
| Antigen | CXCR7/RDC-1 |
|---|---|
| Clone | 11G8 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18731169
![]() |
R&D Systems
FAB4227P-025 |
25 Tests |
251.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18758447
![]() |
R&D Systems
FAB4227P-100 |
100 Tests |
603.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCR7/RDC-1 Monoclonal specifically detects CXCR7/RDC-1 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| CXCR7/RDC-1 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Human, Mouse | |
| ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog | |
| CXCR7 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human CXCR7/RDC-1 in flow cytometry of five distinct human CXCR7 transfectants, but not their respective parental lines. |
| 11G8 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| P25106 | |
| 57007 | |
| Human CXCR7 encoding plasmid Accession # P25106 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title