missing translation for 'onlineSavingsMsg'
Learn More

DLP1 Mouse, Unlabeled, Clone: 8, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611113

 View more versions of this product

Product Code. 15875799

  • ONLINE EXCLUSIVE PRICE
    336.00 €
    323.14 € / 1 set
    Save 12.86 €
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Dynamins, phosphoproteins with intrinsic GTPase activities, play an important role during early endocytosis. The GTPase activity is stimulated in vitro by microtubules, SH3 domain containing-proteins, and Protein Kinase (PKC)- mediated phosphorylation. Although highly related, dynamins are differentially expressed in an organism. For instance, dynamin I is found only in the nervous system, dynamin II is ubiquitously expressed, and dynamin III is located in lung, testis and brain. Due to four alternative splicing products for each of the dynamins, the dynamin family contains at least 12 different members. The Dynamin Like Protein-1 (DLP1) is a new member of the dynamin family of proteins. DLP1 (also known as DVLP) is widely expressed as two alternatively spliced products, each with different tissue expression patterns. The larger product is about 84kDa and reportedly found only in the brain, whereas the 79kDa product is ubiquitously expressed. Unlike dynamins I-III which co-localize with endocytotic vesicles, DLP1 is found aligned with microtubules and with the endoplasmic reticulum, suggesting a secretory function for this novel protein.

Host Species: Mouse
Clone: 8
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat DLP1 aa. 601-722
Formula Weight [Chemical]: 79-84kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DLP1
Monoclonal
250μg/mL
Dynamin Like Protein-1; DVLP
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
8
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Rat DLP1 aa. 601-722
150 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.