missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MICA Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
247.00 € - 546.00 €
Specifications
| Antigen | MICA |
|---|---|
| Clone | 159227 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18788298
![]() |
R&D Systems
FAB1300G-025 |
25 Tests |
247.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18778347
![]() |
R&D Systems
FAB1300G-100 |
100 Tests |
546.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MICA Monoclonal specifically detects MICA in Human samples. It is validated for Flow Cytometry.Specifications
| MICA | |
| Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 100507436 | |
| BaF3 mouse pro-B cell line transfected with human MICA | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 159227 | |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Mouse | |
| Human | |
| FLJ60820, MGC111087, PERB11.1 | |
| MICA | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human MICA in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human MICB. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title