missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-BI Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
316.00 € - 627.00 €
Specifications
| Classification | Polyclonal |
|---|---|
| Target Species | Human, Mouse |
| Gene Accession No. | Q61009 |
| Gene Symbols | SCARB1 |
| Content And Storage | Store at 4C. Do not freeze. |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18787253
|
Novus Biologicals
NB400-104SS |
316.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18774933
|
Novus Biologicals
NB400-104 |
0.1 mL |
627.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SR-BI Polyclonal specifically detects SR-BI in Human, Mouse, Rat, Porcine, Chinese Hamster, Hamster, Mustelid, Primate, Rabbit, Golden Syrian Hamster samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, Knockout Validated, Knockdown Validated, Block/Neutralize.Specifications
| Polyclonal | |
| Q61009 | |
| Store at 4C. Do not freeze. |
| Human, Mouse | |
| SCARB1 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title