missing translation for 'onlineSavingsMsg'
Learn More

AP3B1 Antibody [mFluor Violet 610 SE], Novus Biologicals Biologicals™

Product Code. 30510811 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30510811 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30510811 Supplier Novus Biologicals Supplier No. NBP338363MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AP3B1 Polyclonal antibody specifically detects AP3B1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP3B1
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias Adapter-related protein complex 3 subunit beta-1, Adaptor protein complex AP-3 subunit beta-1, adaptor-related protein complex 3, beta 1 subunit, ADTB3, ADTB3APE, AP-3 complex subunit beta-1, beta-3A-adaptin, beta3A-adaptin, Clathrin assembly protein complex 3 beta-1 large chain, HPS, HPS2AP-3 complex beta-3A subunit
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 895-1094 of human AP3B1 (NP_003655.3).,, Sequence:, FGDKMVSIQITLNNTTDRKIENIHIGEKKLPIGMKMHVFNPIDSLEPEGSITVSMGIDFCDSTQTASFQLCTKDDCFNVNIQPPVGELLLPVAMSEKDFKKEQGVLTGMNETSAVIIAAPQNFTPSVIFQKVVNVANVGAVPSGQDNIHRFAAKTVHSGSLMLVTVELKEGSTAQLIINTEKTVIGSVLLRELKPVLSQG
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8546
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.