missing translation for 'onlineSavingsMsg'
Learn More

APC Antibody (ALi 12-28), HRP, R&D Systems™

Product Code. 18620079 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18620079 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18620079 Supplier Novus Biologicals Supplier No. NBP308922H
  • ONLINE EXCLUSIVE PRICE
    694.69 €
    607.95 € / 100µL
    Save 86.74 €
    12% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

APC Monoclonal specifically detects APC in Human∣Mouse (Negative) samples. It is validated for ELISA∣Flow Cytometry∣Immunocytochemistry/Immunofluorescence∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APC
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone ALi 12-28
Conjugate HRP
Formulation PBS
Gene Alias adenomatosis polyposis coli, adenomatosis polyposis coli tumor suppressor, adenomatous polyposis coli, adenomatous polyposis coli protein, BTPS2, Deleted in polyposis 2.5, DP2.5DP2, DP3, FPC, GS, Protein APC
Host Species Mouse
Immunogen Recombinant fragment of human APC protein (around aa 1-433) (Uniprot: P25054)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Metastasis, Tumor Suppressors, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 324
Target Species Human, Mouse (Negative)
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.