missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APLP-2 Antibody (S06-8I7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19833-100ul
This item is not returnable.
View return policy
Description
APLP-2 Monoclonal antibody specifically detects APLP-2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| APLP-2 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S06-8I7 | |
| Unconjugated | |
| amyloid beta (A4) precursor-like protein 2, amyloid precursor protein homolog HSD-2, Amyloid protein homolog, amyloid-like protein 2, APLP-2, APPHCDEI box-binding protein, APPL2, CDEBP | |
| A synthetic peptide of human APLP-2 (Uniprot # Q06481) | |
| 100 μg | |
| Signal Transduction | |
| 334 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction