missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APLP-2 Antibody (S06-8I7), Novus Biologicals™
Rabbit Monoclonal Antibody
444.00 € - 680.00 €
Specifications
| Antigen | APLP-2 |
|---|---|
| Clone | S06-8I7 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18674943
|
Novus Biologicals
NBP3-19833-50ul |
50 μg |
444.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648453
|
Novus Biologicals
NBP3-19833-100ul |
100 μg |
680.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APLP-2 Monoclonal antibody specifically detects APLP-2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| APLP-2 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| amyloid beta (A4) precursor-like protein 2, amyloid precursor protein homolog HSD-2, Amyloid protein homolog, amyloid-like protein 2, APLP-2, APPHCDEI box-binding protein, APPL2, CDEBP | |
| A synthetic peptide of human APLP-2 (Uniprot # Q06481) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S06-8I7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 334 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title