missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein A-IV/ApoA4 Antibody (CL0468) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Apolipoprotein A-IV/ApoA4 Monoclonal antibody specifically detects Apolipoprotein A-IV/ApoA4 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
Specifications
Specifications
| Antigen | Apolipoprotein A-IV/ApoA4 |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry |
| Classification | Monoclonal |
| Clone | CL0468 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulation | Lyophilized in 5% Trehalose. |
| Gene Alias | apo-AIV, ApoA-IV, Apolipoprotein A4, apolipoprotein A-IV, MGC142154, MGC142156 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?