missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Apolipoprotein A5 Monoclonal specifically detects Apolipoprotein A5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (Negative), CyTOF-ready.
Specifications
Specifications
| Antigen | Apolipoprotein A5 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1G5G9 |
| Concentration | 1.9 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 ug/ml, Flow Cytometry 1 ug per million cells, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, ICC/IF (Negative), CyTOF-ready |
| Gene Accession No. | Q6Q788 |
| Gene Alias | Apo-AV, ApoA-V, APOAV, Apolipoprotein A5, apolipoprotein A-V, FLJ97995, MGC126836, RAP3MGC126838, Regeneration-associated protein 3 |
| Gene Symbols | APOA5 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?