missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Apolipoprotein H/ApoH Antibody (APOH/3682), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26807-20ug
This item is not returnable.
View return policy
Description
Apolipoprotein H/ApoH Monoclonal antibody specifically detects Apolipoprotein H/ApoH in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Apolipoprotein H/ApoH | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Activated protein C-binding protein, Anticardiolipin cofactor, APC inhibitor, Apo-H, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, Beta(2)GPI, beta-2-glycoprotein 1, Beta-2-glycoprotein I, BG | |
| Recombinant fragment (around aa1-300) of human Apolipoprotein H/ApoH protein (exact sequence is proprietary) | |
| 20 μg | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 350 | |
| Store at 4°C. | |
| IgG2c κ |
| Immunohistochemistry (Paraffin) | |
| APOH/3682 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction