missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Apolipoprotein H/ApoH Antibody (APOH/3682), Novus Biologicals™
Mouse Monoclonal Antibody
232.00 € - 462.00 €
Specifications
| Antigen | Apolipoprotein H/ApoH |
|---|---|
| Clone | APOH/3682 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043967
|
Novus Biologicals™
NBP3-26807-100ug |
100 μg |
462.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043285
|
Novus Biologicals™
NBP3-26807-20ug |
20 μg |
232.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Apolipoprotein H/ApoH Monoclonal antibody specifically detects Apolipoprotein H/ApoH in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Apolipoprotein H/ApoH | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 10 mM PBS with 0.05% BSA | |
| 350 | |
| IgG2c κ | |
| Protein A or G purified |
| APOH/3682 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Activated protein C-binding protein, Anticardiolipin cofactor, APC inhibitor, Apo-H, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, Beta(2)GPI, beta-2-glycoprotein 1, Beta-2-glycoprotein I, BG | |
| Recombinant fragment (around aa1-300) of human Apolipoprotein H/ApoH protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title