missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein L1 Antibody (5E2) [DyLight 680], Novus Biologicals Biologicals™

Product Code. 30518361 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30518361 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30518361 Supplier Novus Biologicals Supplier No. NBP342929FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Apolipoprotein L1 Monoclonal antibody specifically detects Apolipoprotein L1 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein L1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 500
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias ApoL, APO-L, ApoL-I, Apolipoprotein L, apolipoprotein L, 1, apolipoprotein L1, Apolipoprotein L-I, FSGS4
Host Species Mouse
Immunogen apolipoprotein L, 1 - (Uniprot# O14791)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 8542
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.