missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein L1 Antibody (CL0170) [DyLight 350], Novus Biologicals™

Product Code. 30297204 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30297204 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30297204

Brand: Novus Biologicals NBP252870UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Apolipoprotein L1 Monoclonal antibody specifically detects Apolipoprotein L1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein L1
Applications Western Blot
Classification Monoclonal
Clone CL0170
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias ApoL, APO-L, ApoL-I, Apolipoprotein L, apolipoprotein L, 1, apolipoprotein L1, Apolipoprotein L-I, FSGS4
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from O14791, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 8542
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.