missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein M/ApoM Antibody (8F12C6B8) [Janelia Fluor« 646], Novus Biologicals Biologicals™

Product Code. 30503721 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30503721 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30503721 Supplier Novus Biologicals Supplier No. NBP327911JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Apolipoprotein M/ApoM Monoclonal antibody specifically detects Apolipoprotein M/ApoM in Human samples. It is validated for ELISA,Western Blot,Serum
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein M/ApoM
Applications ELISA, Western Blot
Classification Monoclonal
Clone 8F12C6B8
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias apolipoprotein M, apo-M, ApoM, G3A, HSPC336, MGC22400, NG20, NG20-like protein, Protein G3a
Host Species Mouse
Immunogen Full-length recombinant Apolipoprotein M/ApoM expressed in E. Coli strain BL21 (DE3).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55937
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1, IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.