missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30865-0.025ML
This item is not returnable.
View return policy
Description
Aquaporin-9 Polyclonal specifically detects Aquaporin-9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Aquaporin-9 | |
| Polyclonal | |
| Unconjugated | |
| Whole antisera with No Preservative | |
| AQP-9, Aquaglyceroporin-9, aquaporin 9, aquaporin-9, HsT17287, Small solute channel 1, SSC1aquaglyceroporin-9 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| 366 | |
| Reconstitute with 0.1 ml sterilized water to desired concentration. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| LYOPH | |
| Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:10-1:500 | |
| O43315 | |
| AQP9 | |
| A synthetic peptide from the c-terminal region of human Aquaporin 9 (AQP9) conjugated to an immunogenic carrier protein was used as the antigen. | |
| 0.025 mL | |
| Primary | |
| Human. Other species not yet tested. | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction