missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
180.00 € - 509.00 €
Specifications
| Antigen | Aquaporin-9 |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18271855
|
Novus Biologicals
NBP1-30865-0.025ML |
0.025 mL |
180.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18798873
|
Novus Biologicals
NBP1-30865 |
509.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Aquaporin-9 Polyclonal specifically detects Aquaporin-9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Aquaporin-9 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| AQP-9, Aquaglyceroporin-9, aquaporin 9, aquaporin-9, HsT17287, Small solute channel 1, SSC1aquaglyceroporin-9 | |
| AQP9 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| O43315 | |
| 366 | |
| IgG | |
| Unpurified | |
| Human. Other species not yet tested. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title