missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ARF5 Monoclonal antibody specifically detects ARF5 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry
Specifications
Specifications
| Antigen | ARF5 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Clone | SR1282 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | ADP-ribosylation factor 5 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human ARF5 (Uniprot #: P84085) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?