missing translation for 'onlineSavingsMsg'
Learn More

ARHGEF5 Antibody, R&D Systems™

Product Code. 18608265 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18608265 25 μL 25µL
18622966 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18608265 Supplier Novus Biologicals Supplier No. NBP24670125ul
  • ONLINE EXCLUSIVE PRICE
    459.38 €
    391.65 € / 25µL
    Save 67.73 €
    14% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

ARHGEF5 Polyclonal specifically detects ARHGEF5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF5
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2), 40% Glycerol
Gene Accession No. A5YM69
Gene Alias ephexin-3, GEF5, Guanine nucleotide regulatory protein TIM, Oncogene TIM, P60, p60 TIM, Rho guanine nucleotide exchange factor (GEF) 5, rho guanine nucleotide exchange factor 5, TIM1, TIMDKFZp686N1969, Transforming immortalized mammary oncogene
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: QGEERKRELQVPEENRADSQDEKSQTFLGKSEEVTGKQEDHGIKEKGVPVSGQEAKEPESWDGGRLGAVGRARSREEENEHHGPSMPALIAPEDSPHCDLFP
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7984
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.