missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aripiprazole metabolite (OPC-3373), 1mg/ml in Ethanol, Lipomed
Description
Aripiprazole metabolite (OPC-3373), 1mg/ml in Ethanol
Specifications
Specifications
| Chemical Name or Material | 3,4-Dihydro-7-(3'carboxy)propoxy-2(1 H)-quinolinone |
| CAS | 58899-27-7 |
| Analyte or Component Names | 3,4-Dihydro-7-(3'carboxy)propoxy-2(1 H)-quinolinone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H15 N O4 |
| Concentration | 1 mg/mL |
| Synonym | Butanoic acid, 4-[(1,2,3,4-tetrahydro-2-oxo-7-quinolinyl)oxy]-, 4-[(1,2,3,4-Tetrahydro-2-oxo-7-quinolinyl)oxy]butanoic acid, 4-[(2-Oxo-3,4-dihydroquinolin-7(1 H)-yl)oxy]butanoic Acid |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | OC(=O)CCCOc1ccc2CCC(=O)Nc2c1 |
| IUPAC Name | 4-[(2-oxo-3,4-dihydro-1 H-quinolin-7-yl)oxy]butanoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?