missing translation for 'onlineSavingsMsg'
Learn More

Aristolochic acid I, 96%

Product Code. p-3777340 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10420501 100 mg Glass bottle 100mg
10005913 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10420501 Supplier Thermo Scientific Acros Supplier No. 226091000
Add to basket
This item is not returnable. View return policy

CAS: 313-67-7 | C17H11NO7 | 341.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313-67-7
Molecular Formula C17H11NO7
Molecular Weight (g/mol) 341.27
MDL Number MFCD00004996
InChI Key BBFQZRXNYIEMAW-UHFFFAOYSA-N
Synonym aristolochic acid, aristolochic acid a, aristolochic acid i, aristolochin, tardolyt, birthwort, aristolochic acid-i, aristolochine, aristolochiazaeure, unii-94218wfp5t
PubChem CID 2236
ChEBI CHEBI:2825
SMILES COC1=CC=CC2=C3C(=C(C=C21)[N+](=O)[O-])C(=CC4=C3OCO4)C(=O)O

Specifications

Melting Point 269°C to 270°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (typically I+ II isomers) (HPLC)
Packaging Glass bottle
Quantity 100 mg
Beilstein 19, V,7, 611
Merck Index 15, 777
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,chloroform,ether,acetone,,acetic acid,aniline,alkalies,practically insoluble in benzene,carbon disulfide
Formula Weight 341.27
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Aristolochic acid, Mixture of I and II
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aristolochic acid, mixture of I and II
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1A
Hazard Statement
  • H301-Toxic if swallowed.
  • H350-May cause cancer.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.