missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPC3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92679-0.02ml
This item is not returnable.
View return policy
Description
ARPC3 Polyclonal antibody specifically detects ARPC3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| ARPC3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| actin related protein 2/3 complex, subunit 3, 21kDa, subunit 3 (21 kD) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-178 of human ARPC3 (NP_001265485.1). MPAYHSSLMDPDTKLIGNMALLPIRSQFKGPAPRETKDTDIVDEAIYYFKANVFFKNYEIKNEADRTLIYITLYISECLKKLQKCNSKSQGEKEMYTLGITNFPIPGEPGFPLNAIYAKPANKQEDEVMRAYLQQLRQETGLRLCEKVFDPQNDKPSKWWTCFVKRQFMNKSLSGPGQ | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 10094 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction