missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPC3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 463.00 €
Specifications
| Antigen | ARPC3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661122
|
Novus Biologicals
NBP2-92679-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686142
|
Novus Biologicals
NBP2-92679-0.1ml |
0.1 mL |
463.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARPC3 Polyclonal antibody specifically detects ARPC3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| ARPC3 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 10094 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| actin related protein 2/3 complex, subunit 3, 21kDa, subunit 3 (21 kD) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-178 of human ARPC3 (NP_001265485.1). MPAYHSSLMDPDTKLIGNMALLPIRSQFKGPAPRETKDTDIVDEAIYYFKANVFFKNYEIKNEADRTLIYITLYISECLKKLQKCNSKSQGEKEMYTLGITNFPIPGEPGFPLNAIYAKPANKQEDEVMRAYLQQLRQETGLRLCEKVFDPQNDKPSKWWTCFVKRQFMNKSLSGPGQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title