Learn More
RETSCH AS 200 Standard Clamping Devices
Product Code. 10136791 Shop All Retsch Products
Change view
Click to view available options
For Use With (Application):
Dry Sieving
Wet Sieving
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
10136791 Dry Sieving Each
10789902 Wet Sieving Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10136791 Supplier RETSCH Supplier No. 32.662.0005
  • ONLINE EXCLUSIVE PRICE
    915.00 €
    844.29 € / Pack of 1
    Save 70.71 €
    7% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Clamping Devices
Diameter (Metric) 100/150/200/203 mm
For Use With (Application) Dry Sieving
For Use With (Equipment) Analytical sieve shaker AS 200
Packaging Quantity 1 Ea.