missing translation for 'onlineSavingsMsg'
Learn More

ASZ1 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18628632 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18628632 0.02 mL 0.02mL
18649511 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18628632 Supplier Novus Biologicals Supplier No. NBP2928630.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ASZ1 Polyclonal antibody specifically detects ASZ1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASZ1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ALP1Germ cell-specific ankyrin, SAM and basic leucine zipper domain-containingprotein, ANKL14933400N19Rik, ankyrin repeat, SAM and basic leucine zipper domain containing 1, ankyrin repeat, SAM and basic leucine zipper domain-containing protein 1, ankyrin-like 1, Ankyrin-like protein 1, CT1.19, GASZC7orf7, MGC26634, Orf3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of human ASZ1 (NP_570124.1). MAASALRGLPVAGGGESSESEDDGWEIGYLDRTSQKLKRLLPIEEKKEKFKKAMTIGDVSLVQELLDSGISVDSNFQYGWTPLMYAASVANAELVRVLLDRGANASFEKDKQSILITACSAHGSEEQILKCVELLLSRNADPNVACRRLMTPIMYAARDGHTQVVALLVAHGAEVNTQDENGYTALTWAARQGHKNIVLKLLELGANKMLQTKDGKMPSEIAKRNKHHEIFNLLSFTLNPLEGKLQQLTKEDTICKILTTDSDREKDHIFSSYTAFGDLEVFLHGLGLEHMTDLLKERDITLRHLLTMREDEFTKNGITSKDQQKILAALKELQVEEIQFGELSEETKLE
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Endocrinology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 136991
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.