missing translation for 'onlineSavingsMsg'
Learn More

Atenolol, Lipomed

Product Code. 30460425
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460425 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460425 Supplier Lipomed Supplier No. LPMATN1853FB10
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Atenolol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Atenolol
CAS 29122-68-7
Analyte or Component Names Atenolol
Shipping Condition Room Temperature
Molecular Formula C14 H22 N2 O3
Synonym Benzeneacetamide, 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-, Acetamide, 2-[p-[2-hydroxy-3-(isopropylamino)propoxy]phenyl]- (8 CI), (RS)-Atenolol, Alinor, Altol, Anselol, Antipressan, Apo-Atenolol, Atcardil, AteHexal, Atecard, Atelol, Aten, Atenblock, Atendol, Atenet, Ateni, Atenil, Ateno, Atenol, Atenolol, Atereal, Aterol, Atolmin, B-Vasc, Betablok, Betacard, Blokium, Catenol, Coratol, Corotenol, Cuxanorm, DL-Atenolol, Duraatenolol, Evitocor, Farnormin, Felo-Bits, Hipres, Hypoten, ICI 66082, Ibinolo, Internolol, Lo-ten, Lotenal, Myocord, Normalol, Normiten, Noten, Novaten, Oraday, Premorine, Prenolol, Prenormine, Seles, Seles Beta, Selobloc, Serten, Stermin, Telol, Tenidon, Teno-basan, Tenoblock, Tenolin, Tenolol, Tenoprin, Tenordin, Tenormin, Tenormine, Tensig, Tensimin, Ternomin, Tredol, Uniloc, Urosin, Vascoten, Vericordin, Wesipin, Xaten, dl-Atenolol, (+/-)-Atenolol
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CC(C)NCC(O)COc1ccc(CC(=O)N)cc1
IUPAC Name 2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 266.34
Quantity 10 mg
Formula Weight 266.163 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.