missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-02935SS
This item is not returnable.
View return policy
Description
ATF3 Polyclonal specifically detects ATF3 in Human, Mouse, Rat, Mammal samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ATF3 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| activating transcription factor 3cAMP-dependent transcription factor ATF-3, ATF3deltaZip2, ATF3deltaZip2c, ATF3deltaZip3, cyclic AMP-dependent transcription factor ATF-3 | |
| Rabbit | |
| 17 kDa | |
| 0.025 mL | |
| Cancer, Transcription Factors and Regulators | |
| 467 | |
| Human, Mouse, Rat, Mammalia | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.27 mg/mL | |
| Western Blot 1:200, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| P18847 | |
| ATF3 | |
| A synthetic peptide corresponding to a region of human ATF3 (within residues 100-150). [UniProt# P18847] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse and rat. Predicted to react with most mammalian species based on sequence homology. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction