missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
268.00 € - 527.00 €
Specifications
| Antigen | ATF3 |
|---|---|
| Concentration | 1.27 mg/mL |
| Dilution | Western Blot 1:200, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|
18798223
|
Novus Biologicals
NBP1-02935SS |
0.025 mL |
268.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18760434
|
Novus Biologicals
NBP1-02935 |
0.1 mL |
527.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
ATF3 Polyclonal specifically detects ATF3 in Human, Mouse, Rat, Mammal samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifikationer
| ATF3 | |
| Western Blot 1:200, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Cancer, Transcription Factors and Regulators | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| activating transcription factor 3cAMP-dependent transcription factor ATF-3, ATF3deltaZip2, ATF3deltaZip2c, ATF3deltaZip3, cyclic AMP-dependent transcription factor ATF-3 | |
| ATF3 | |
| IgG | |
| Affinity Purified | |
| Human, mouse and rat. Predicted to react with most mammalian species based on sequence homology. |
| 1.27 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Mammalia | |
| P18847 | |
| 467 | |
| A synthetic peptide corresponding to a region of human ATF3 (within residues 100-150). [UniProt# P18847] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 17 kDa |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel