missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24594
This item is not returnable.
View return policy
Description
ATG10 Polyclonal specifically detects ATG10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ATG10 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| APG10, APG10 autophagy 10-like (S. cerevisiae), APG10L, APG10-like, ATG10 autophagy related 10 homolog (S. cerevisiae), Autophagy-related protein 10, DKFZp586I0418, EC 6.3.2.-, FLJ13954, pp12616, ubiquitin-like-conjugating enzyme ATG10 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoform 2, isoform 3, isoform 4, isoform 5, isoform 6 and isoform 7) and 92% homologous in mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 2.0-6.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Q9H0Y0 | |
| ATG10 | |
| A portion of amino acids 1-50 of human APG10L was used as the immunogen. | |
| 0.1 mg | |
| Autophagy | |
| 83734 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction