missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
199.00 € - 351.00 €
Specifications
| Antigen | ATG10 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2.0-6.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18715913
|
Novus Biologicals
NBP2-24594SS |
0.025 mg |
199.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18167001
|
Novus Biologicals
NBP2-24594 |
0.1 mg |
351.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG10 Polyclonal specifically detects ATG10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ATG10 | |
| Western Blot 2.0-6.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Q9H0Y0 | |
| 83734 | |
| A portion of amino acids 1-50 of human APG10L was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Autophagy | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| APG10, APG10 autophagy 10-like (S. cerevisiae), APG10L, APG10-like, ATG10 autophagy related 10 homolog (S. cerevisiae), Autophagy-related protein 10, DKFZp586I0418, EC 6.3.2.-, FLJ13954, pp12616, ubiquitin-like-conjugating enzyme ATG10 | |
| ATG10 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoform 2, isoform 3, isoform 4, isoform 5, isoform 6 and isoform 7) and 92% homologous in mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title