missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG16L1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 9 publications
Brand: Novus Biologicals NB110-60928SS
This item is not returnable.
View return policy
Description
ATG16L1 Polyclonal specifically detects ATG16L1 in Human, Mouse, Rat, Alligator, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Electron Microscopy.
Specifications
| ATG16L1 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| APG16 autophagy 16-like (S. cerevisiae), APG16L beta, APG16LFLJ10828, APG16-like 1, ATG16 autophagy related 16-like (S. cerevisiae), ATG16 autophagy related 16-like 1 (S. cerevisiae), ATG16A, ATG16L, autophagy-related protein 16-1, FLJ00045, FLJ10035, FLJ22677, IBD10, WD repeat domain 30, WDR30 | |
| Rabbit | |
| 68 kDa | |
| 0.025 mL | |
| Autophagy, Cancer, Cell Biology, Macroautophagy, Neurodegeneration, Neuroscience, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
| 55054 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay | |
| 0.8 mg/mL | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:250 - 1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 25060858), Electron Microscopy 1:10-1:500. Use reported in scientific literature (PMID 22531915) | |
| Q676U5 | |
| ATG16L1 | |
| A synthetic peptide within residues 1-100 of human ATG16L1 protein. [Swiss-Prot# Q676U5] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Alligator, Bovine, Canine, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction