missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG16L1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 9 publications
237.00 € - 473.00 €
Specifications
| Antigen | ATG16L1 |
|---|---|
| Concentration | 0.8 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:250 - 1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 25060858), Electron Microscopy 1:10-1:500. Use reported in scientific literature (PMID 22531915) |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18771334
|
Novus Biologicals
NB110-60928SS |
0.025 mL |
237.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18737103
|
Novus Biologicals
NB110-60928 |
0.1 mL |
473.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG16L1 Polyclonal specifically detects ATG16L1 in Human, Mouse, Rat, Alligator, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Electron Microscopy.Specifications
| ATG16L1 | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:250 - 1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 25060858), Electron Microscopy 1:10-1:500. Use reported in scientific literature (PMID 22531915) | |
| Polyclonal | |
| Rabbit | |
| Autophagy, Cancer, Cell Biology, Macroautophagy, Neurodegeneration, Neuroscience, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| APG16 autophagy 16-like (S. cerevisiae), APG16L beta, APG16LFLJ10828, APG16-like 1, ATG16 autophagy related 16-like (S. cerevisiae), ATG16 autophagy related 16-like 1 (S. cerevisiae), ATG16A, ATG16L, autophagy-related protein 16-1, FLJ00045, FLJ10035, FLJ22677, IBD10, WD repeat domain 30, WDR30 | |
| ATG16L1 | |
| IgG | |
| Affinity Purified | |
| 68 kDa |
| 0.8 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Alligator, Bovine, Canine, Primate | |
| Q676U5 | |
| 55054 | |
| A synthetic peptide within residues 1-100 of human ATG16L1 protein. [Swiss-Prot# Q676U5] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title