missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG16L2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92198-0.02ml
This item is not returnable.
View return policy
Description
ATG16L2 Polyclonal antibody specifically detects ATG16L2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ATG16L2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| APG16-like 2, ATG16 autophagy related 16-like 2 (S. cerevisiae), ATG16B, autophagy-related protein 16-2, FLJ00012, WDR80WD repeat-containing protein 80 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-275 of human ATG16L2 (NP_203746.1). MAGPGVPGAPAARWKRHIVRQLRLRDRTQKALFLELVPAYNHLLEKAELLDKFSKKLQPEPNSVTPTTHQGPWEESELDSDQVPSLVALRVKWQEEEEGLRLVCGEMAYQVVEKGAALGTLESELQQRQSRLAALEARVAQLREARAQQAQQVEEWRAQNAVQRAAYEALRAHVGLREAALRRLQEEARDLLERLVQRKARAAAERNLRNERRERAKQARVSQELKKAAKRTVSISEGPDTLGDGMRERRETLALAPEPEPLEKEACEKWKRPFR | |
| 0.02 mL | |
| Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Signal Transduction | |
| 89849 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction