missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG16L2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 463.00 €
Specifications
| Antigen | ATG16L2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627661
|
Novus Biologicals
NBP2-92198-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603801
|
Novus Biologicals
NBP2-92198-0.1ml |
0.1 mL |
463.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG16L2 Polyclonal antibody specifically detects ATG16L2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ATG16L2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 89849 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| APG16-like 2, ATG16 autophagy related 16-like 2 (S. cerevisiae), ATG16B, autophagy-related protein 16-2, FLJ00012, WDR80WD repeat-containing protein 80 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-275 of human ATG16L2 (NP_203746.1). MAGPGVPGAPAARWKRHIVRQLRLRDRTQKALFLELVPAYNHLLEKAELLDKFSKKLQPEPNSVTPTTHQGPWEESELDSDQVPSLVALRVKWQEEEEGLRLVCGEMAYQVVEKGAALGTLESELQQRQSRLAALEARVAQLREARAQQAQQVEEWRAQNAVQRAAYEALRAHVGLREAALRRLQEEARDLLERLVQRKARAAAERNLRNERRERAKQARVSQELKKAAKRTVSISEGPDTLGDGMRERRETLALAPEPEPLEKEACEKWKRPFR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title