missing translation for 'onlineSavingsMsg'
Learn More

ATG4D Antibody [Biotin], Novus Biologicals Biologicals™

Product Code. 30502406 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30502406 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30502406 Supplier Novus Biologicals Supplier No. NBP337970B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATG4D Polyclonal antibody specifically detects ATG4D in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG4D
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias APG4 autophagy 4 homolog D, APG4 autophagy 4 homolog D (S. cerevisiae), APG4D, APG4-D, ATG4 autophagy related 4 homolog D (S. cerevisiae), AUTL4, AUT-like 4 cysteine endopeptidase, AUT-like 4, cysteine endopeptidase, AUT-like 4, cysteine endopeptidase (S. cerevisiae), Autophagin-4, Autophagy-related cysteine endopeptidase 4, Autophagy-related protein 4 homolog D, cysteine protease ATG4D, cysteine protease involved in autophagy, EC 3.4.22, EC 3.4.22.-
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human ATG4D (NP_116274.3).,, Sequence:, MNSVSPAAAQYRSSSPEDARRRPEARRPRGPRGPDPNGLGPSGASGPALGSPGAGPSEPDEVDKFKAKFLTAWNNVKYGWVVKSRTSFSKISSIHLCGRRYRFEGEGDIQRFQRDFVSRLWLTYRRDFPPLPGGCLTSDCGWGCMLRSGQMMLAQGLLLHFLPRDWTWAEGMGLGPPELSGSASPSRYHGPARWMPPRWAQGAPELEQERRHRQIVSWFADHPRAPFGLH
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 84971
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.