missing translation for 'onlineSavingsMsg'
Learn More

ATP12A Antibody, Novus Biologicals™

Product Code. p-200059632 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18699548 100 μg 100µL
18688298 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18699548 Supplier Novus Biologicals Supplier No. NBP306153100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP12A Polyclonal antibody specifically detects ATP12A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP12A
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:400
Formulation PBS, pH7.0
Gene Alias ATP1AL1non-gastric H(+)/K(+) ATPase alpha subunit, ATPase, H+/K+ transporting, nongastric, alpha polypeptide, ATPase, Na+/K+ transporting, alpha polypeptide-like 1, EC 3.6.3, EC 3.6.3.10, Non-gastric H(+)/K(+) ATPase subunit alpha, potassium-transporting ATPase alpha chain 2, Proton pump
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ATP12A.
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 479
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.