Learn More
ATP1A1-Specific Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-55187-100UL
Description
The ATP1A1-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human ATP1A1-Specific. This antibody recognizes human, mouse antigen. The ATP1A1-Specific antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| ATP1A1-Specific | |
| 1000 μg/mL | |
| ATP1A1, Sodium pump subunit alpha 1 | |
| 100 μL | |
| 476 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |