Learn More
ATP1A1-Specific Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 450.00 €
Specifications
| Antigen | ATP1A1-Specific |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18088045
|
Proteintech
CL488-55187-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18050706
|
Proteintech
CL488-55187-100UL |
100 μL |
450.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ATP1A1-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human ATP1A1-Specific. This antibody recognizes human, mouse antigen. The ATP1A1-Specific antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| ATP1A1-Specific | |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse | |
| ATP1A1, Sodium pump subunit alpha 1 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 476 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.