Learn More
ATP1A1-Specific Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-55187-20UL
 View more versions of this product
Product Code. 18088045
  • 145.00 € / 20µL
Description
The ATP1A1-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human ATP1A1-Specific. This antibody recognizes human, mouse antigen. The ATP1A1-Specific antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
ATP1A1-Specific
1000 μg/mL
ATP1A1, Sodium pump subunit alpha 1
20 μL
476
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Primary
Human, Mouse
150 kDa