missing translation for 'onlineSavingsMsg'
Learn More

ATP6V1E1 Antibody [mFluor Violet 450 SE], Novus Biologicals Biologicals™

Product Code. 30497616 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30497616 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30497616 Supplier Novus Biologicals Supplier No. NBP338049MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP6V1E1 Polyclonal antibody specifically detects ATP6V1E1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V1E1
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias ATP6E2ATP6V1E, ATP6E31kDa subunit, ATPase, H+ transporting, lysosomal (vacuolar proton pump) 31kD, ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E isoform 1, ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E1, H+-transporting ATP synthase chain E, vacuolar, p31, Vacuolar proton pump subunit E 1, V-ATPase 31 kDa subunit, V-ATPase subunit E 1, V-ATPase, subunit E, Vma4, V-type proton ATPase subunit E 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 77-226 of human ATP6V1E1 (NP_001687.1).,, Sequence:, NQARLKVLRARDDLITDLLNEAKQRLSKVVKDTTRYQVLLDGLVLQGLYQLLEPRMIVRCRKQDFPLVKAAVQKAIPMYKIATKNDVDVQIDQESYLPEDIAGGVEIYNGDRKIKVSNTLESRLDLIAQQMMPEVRGALFGANANRKFLD
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 529
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.