missing translation for 'onlineSavingsMsg'
Learn More

ATPG Antibody (8H11) [Janelia Fluor« 549], Novus Biologicals Biologicals™

Product Code. 30518426 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30518426 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30518426 Supplier Novus Biologicals Supplier No. NBP342931JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ATPG Monoclonal antibody specifically detects ATPG in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATPG
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8H11
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias ATP synthase gamma chain, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1, ATP5CATP synthase subunit gamma, mitochondrial, ATP5CL1mitochondrial ATP synthase, gamma subunit 1, F-ATPase gamma subunit
Host Species Mouse
Immunogen ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1 - (Uniprot# P36542)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 509
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.