missing translation for 'onlineSavingsMsg'
Learn More

Atrial Natriuretic Peptide/ANP Antibody [Janelia Fluor« 549], Novus Biologicals Biologicals™

Product Code. 30499520 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30499520 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30499520 Supplier Novus Biologicals Supplier No. NBP335467JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Atrial Natriuretic Peptide/ANP Polyclonal antibody specifically detects Atrial Natriuretic Peptide/ANP in Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Atrial Natriuretic Peptide/ANP
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias ANF, ANPatriopeptin, ATFB6, atrial natriuretic factor, Atrial natriuretic peptide, CDD-ANF, natriuretic peptide A, natriuretic peptide precursor A, PNDcardionatrin, prepronatriodilatin
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 26-151 of human Atrial Natriuretic Peptide/ANP (NP_006154.1).,, Sequence:, LTWQEIMSITELQGLNAPSEPSFEPQAPAPYLGPPPPTTYCPCSIHPDSGFPLPPPPYELPASTSHVPDPPYSYGNMAIPVSKPLSLSGLLSEPLQDPLALLDIGLPAGPPKPQEDPESDSGLSLN
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4878
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.