missing translation for 'onlineSavingsMsg'
Learn More

ATRX Antibody (CL0537) [DyLight 594], Novus Biologicals™

Product Code. 30525559 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30525559 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30525559 Supplier Novus Biologicals Supplier No. NBP343870DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ATRX Monoclonal antibody specifically detects ATRX in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATRX
Applications Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL0537
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias alpha thalassemia/mental retardation syndrome X-linked, alpha thalassemia/mental retardation syndrome X-linked (RAD54 (S. cerevisiae)homolog), alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog, S.cerevisiae), ATP-dependent helicase ATRX, ATR2, DNA dependent ATPase and helicase, EC 3.6.1, EC 3.6.4.12, helicase 2, X-linked, Juberg-Marsidi syndrome, MGC2094, MRXHF1, RAD54 homolog, RAD54L, SFM1, SHS, transcriptional regulator ATRX, XH2RAD54, X-linked helicase II, X-linked nuclear protein, XNPZNF-HX, Zinc finger helicase, Znf-HX
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of P46100, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 546
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.