missing translation for 'onlineSavingsMsg'
Learn More

Aureusimine A, TRC

Product Code. 30135770
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135770 50 mg 50mg
30010543 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135770 Supplier Toronto Research Chemicals Supplier No. TRCA79456050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1244033-70-2
Molecular Formula C14H16N2O2
Molecular Weight (g/mol) 244.29
Synonym 6-[(4-Hydroxyphenyl)methyl]-3-(1-methylethyl)-2(1H)-pyrazinone, Tyrvalin;
IUPAC Name 6-[(4-hydroxyphenyl)methyl]-3-propan-2-yl-1H-pyrazin-2-one
SMILES CC(C)C1=NC=C(CC2=CC=C(C=C2)O)NC1=O

Specifications

Analyte or Component Names Aureusimine A
Shipping Condition Room Temperature
InChI Formula InChI=1S/C14H16N2O2/c1-9(2)13-14(18)16-11(8-15-13)7-10-3-5-12(17)6-4-10/h3-6,8-9,17H,7H2,1-2H3,(H,16,18)
Solution Type Neat
Industry Type Forensics
Quantity 50 mg
Formula Weight 244.1212
Chemical Name or Material Aureusimine A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.