missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza A H7N9 Hemagglutinin Antibody - (A/blue-winged teal/Ohio/566/2006) - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41276-0.025mg
This item is not returnable.
View return policy
Description
Avian Influenza A H7N9 Hemagglutinin Polyclonal specifically detects Avian Influenza A H7N9 Hemagglutinin in Virus samples. It is validated for ELISA.
Specifications
| Avian Influenza A H7N9 Hemagglutinin | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| virus | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| ELISA | |
| 1 mg/mL | |
| ELISA 1 μg/mL | |
| H7N9HA | |
| Antibody was raised against a synthetic peptide corresponding to 11 amino acids near the center of the hemagglutinin [Influenza A virus (A/blue-winged teal/Ohio/566/2006(H7N9))] protein. The immunogen is located within amino acids 260 - 310 of Avian Influenza A H7N9 Hemagglutinin. Amino Acid Squence: GVQSDVPLDSG | |
| 0.025 mg | |
| Primary | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction