missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza A H7N9 Hemagglutinin Antibody - (A/blue-winged teal/Ohio/566/2006) - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00 € - 507.00 €
Specifications
| Antigen | Avian Influenza A H7N9 Hemagglutinin |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18646128
|
Novus Biologicals
NBP2-41276-0.025mg |
0.025 mg |
168.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18193928
|
Novus Biologicals
NBP2-41276 |
0.1 mg |
507.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Avian Influenza A H7N9 Hemagglutinin Polyclonal specifically detects Avian Influenza A H7N9 Hemagglutinin in Virus samples. It is validated for ELISA.Specifications
| Avian Influenza A H7N9 Hemagglutinin | |
| ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a synthetic peptide corresponding to 11 amino acids near the center of the hemagglutinin [Influenza A virus (A/blue-winged teal/Ohio/566/2006(H7N9))] protein. The immunogen is located within amino acids 260 - 310 of Avian Influenza A H7N9 Hemagglutinin. Amino Acid Squence: GVQSDVPLDSG | |
| Primary | |
| virus |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Virus | |
| H7N9HA | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title