Learn More
Azlon™ Polypropylene Test Tube Rack
Economical and sturdy test tube racks supplied in self assembly flat packs
69.80 € - 116.00 €
Product Code | Brand | Color | Positions | Diameter (Metric) Opening | Dimensions (L x W x H) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Positions | Diameter (Metric) Opening | Dimensions (L x W x H) | Price | Quantity & Availability | ||
11387794
|
Azlon
RPB021P |
Blue | 40 | 21 mm | 265 x 126 x 75mm |
92.40 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
12531406
|
Azlon
RPB026P |
Blue | 32 | 26 mm | 265 x 126 x 88mm |
116.00 €
Pack of 5 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||
11367794
|
Azlon
RPB018P |
Blue | 55 | 18 mm | 265 x 126 x 75mm |
110.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
11933849
|
Azlon
RPB031P |
Blue | 21 | 31 mm | 265 x 126 x 88mm |
116.00 €
Pack of 5 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||
11377794
|
Azlon
RPW021P |
White | 40 | 21 mm | 265 x 126 x 75mm |
103.00 €
Pack of 5 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||
11397794
|
Azlon
RPW026P |
White | 32 | 26 mm | 265 x 126 x 88mm |
104.00 €
Pack of 5 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||
11347794
|
Azlon
RPW013P |
White | 84 | 13 mm | 265 x 126 x 75mm |
92.40 €
Pack of 5 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||
11971528
|
Azlon
RPW018P |
White | 55 | 18 mm | 265 x 126 x 75mm |
103.00 €
Pack of 5 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||
11307804
|
Azlon
RPW031P |
White | 21 | 31 mm | 265 x 126 x 88mm |
116.00 €
Pack of 5 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||
11357794
|
Azlon
RPB013P |
Blue | 84 | 13 mm | 265 x 126 x 75mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Economical and sturdy test tube racks supplied in self assembly flat packs
- Autoclavable at 121°C
- Sinks in water baths
- Alpha numeric place coding
Specifications
Autoclavable | |
Polypropylene |