missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H2/ICOSLG Antibody (599829) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals MAB1581-SP
This item is not returnable.
View return policy
Description
B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| B7-H2/ICOSLG | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry, CyTOF-ready | |
| B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 23308 | |
| Store at -20C in the dark. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 599829 | |
| Unconjugated | |
| PBS with No Preservative | |
| ICOSLG | |
| rmB7-H2 aa 47-279 | |
| 25 μg | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction